Please login to your online account to display your discounted pricing

Armenian Hamster IgG, Clone: 299Arm, APC, Isotype control, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP142999005MG

 View all options for this product

Catalog No. NBP14299905


Description & Specifications


Antigen IgG
Applications Flow Cytometry
Clone 299Arm
Concentration 0.2mg/mL
Conjugate APC
Format Conjugated
Formulation Aqueous buffer, may contain carrier protein/stabilizer (proprietary).
Host Species Armenian Hamster
Isotype IgG
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C in the dark.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity This 299Arm antibody has been tested by flow cytometric analysis of mouse splenocytes and normal human peripheral blood. This should be used at the same concentration as the experimental antibody.