Please login to your online account to display your discounted pricing

Armenian Hamster IgG, Clone: 299Arm, APC, Isotype control, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14299901MG

 View more versions of this product

Catalog No. NBP14299901

  • / Each
 

Description & Specifications

Specifications

Host Species Armenian Hamster
Quantity 0.1mg
Monoclonal or Polyclonal Monoclonal
Applications Flow Cytometry
Antigen IgG
Format Conjugated
Isotype IgG
Storage Requirements Store at 4°C in the dark.
Formulation Aqueous buffer, may contain carrier protein/stabilizer (proprietary).
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This 299Arm antibody has been tested by flow cytometric analysis of mouse splenocytes and normal human peripheral blood. This should be used at the same concentration as the experimental antibody.
Clone 299Arm
Concentration 0.2mg/mL
Conjugate APC