Please login to your online account to display your discounted pricing

Armenian Hamster anti-TRAIL R2/TNFRSF10B, Clone: MD5-1, Novus Biologicals™

$274.00 - $538.00

Specifications

Antigen TRAIL R2/TNFRSF10B
Clone MD5-1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was DR5.
Gene ID (Entrez) 8795
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14328501

 
novus biologicals canada
NBP14328501MG
0.1mg Each for $329.00

NBP14328505

 
novus biologicals canada
NBP143285005MG
0.05mg Each for $274.00

NBP1432855

 
novus biologicals canada
NBP14328505MG
0.5mg Each for $538.00
Description & Specifications

Specifications

Antigen TRAIL R2/TNFRSF10B
Clone MD5-1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was DR5.
Gene ID (Entrez) 8795
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Functional Assay