Please login to your online account to display your discounted pricing

Armenian Hamster anti-TRAIL R2/TNFRSF10B, Clone: MD5-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143285005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14328505

  • / Each
 

Description & Specifications

Specifications

Antigen TRAIL R2/TNFRSF10B
Applications Flow Cytometry
Applications Functional Assay
Clone MD5-1
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Armenian Hamster
Immunogen The immunogen for this antibody was DR5.
Isotype IgG
Purification Method Affinity Purified
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 8795
Target Species Murine