Please login to your online account to display your discounted pricing

Armenian Hamster anti-TRAIL R2/TNFRSF10B, Clone: MD5-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14328505MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1432855

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Armenian Hamster
Applications Flow Cytometry
Applications Functional Assay
Immunogen The immunogen for this antibody was DR5.
Isotype IgG
Storage Requirements Store at 4°C. Do not freeze.
Cross Reactivity Mouse
Antigen TRAIL R2/TNFRSF10B
Quantity 0.5mg
Regulatory Status RUO
Primary or Secondary Primary
Clone MD5-1
Concentration 0.5mg/mL
Purification Method Affinity Purified
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 8795
Conjugate Unlabeled