Please login to your online account to display your discounted pricing

Armenian Hamster anti-TRAIL R2/TNFRSF10B, Clone: MD5-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14328501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14328501

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Mouse
Immunogen The immunogen for this antibody was DR5.
Applications Flow Cytometry
Applications Functional Assay
Monoclonal or Polyclonal Monoclonal
Isotype IgG
Primary or Secondary Primary
Conjugate Unlabeled
Storage Requirements Store at 4°C. Do not freeze.
Clone MD5-1
Antigen TRAIL R2/TNFRSF10B
Formulation Phosphate buffer pH 7.2
Host Species Armenian Hamster
Regulatory Status RUO
Gene ID (Entrez) 8795
Purification Method Affinity Purified
Format Affinity Purified
Concentration 0.5mg/mL
Quantity 0.1mg