Please login to your online account to display your discounted pricing

Armenian Hamster anti-TCR gamma/delta, Clone: GL3, Novus Biologicals™

$219.00 - $362.00

Specifications

Antigen TCR gamma/delta
Clone GL3
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was gamma delta TCR.
Gene ID (Entrez) 6965
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14330701

 
novus biologicals canada
NBP14330701MG
0.1mg Each for $274.00

NBP14330705

 
novus biologicals canada
NBP143307005MG
0.05mg Each for $219.00

NBP1433075

 
novus biologicals canada
NBP14330705MG
0.5mg Each for $362.00
Description & Specifications

Specifications

Antigen TCR gamma/delta
Clone GL3
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was gamma delta TCR.
Gene ID (Entrez) 6965
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Mouse
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunoprecipitation