Please login to your online account to display your discounted pricing

Armenian Hamster anti-TCR gamma/delta, Clone: GL3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143307005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14330705

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Host Species Armenian Hamster
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Gene ID (Entrez) 6965
Conjugate Unlabeled
Cross Reactivity Mouse
Antigen TCR gamma/delta
Formulation Phosphate buffer pH 7.2
Applications Flow Cytometry
Applications Immunoprecipitation
Immunogen The immunogen for this antibody was gamma delta TCR.
Isotype IgG
Purification Method Affinity Purified
Quantity 0.05mg
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Clone GL3
Concentration 0.5mg/mL