Please login to your online account to display your discounted pricing

Armenian Hamster anti-TCR gamma/delta, Clone: GL3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14330701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14330701

  • / Each
 

Description & Specifications

Specifications

Antigen TCR gamma/delta
Formulation Phosphate buffer pH 7.2
Cross Reactivity Mouse
Conjugate Unlabeled
Applications Flow Cytometry
Applications Immunoprecipitation
Quantity 0.1mg
Gene ID (Entrez) 6965
Immunogen The immunogen for this antibody was gamma delta TCR.
Monoclonal or Polyclonal Monoclonal
Format Affinity Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Isotype IgG
Host Species Armenian Hamster
Purification Method Affinity Purified
Primary or Secondary Primary
Clone GL3
Concentration 0.5mg/mL