Please login to your online account to display your discounted pricing

Armenian Hamster anti-TCR gamma/delta, Clone: GL3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14330705MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1433075

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Mouse
Applications Flow Cytometry
Applications Immunoprecipitation
Formulation Phosphate buffer pH 7.2
Regulatory Status RUO
Host Species Armenian Hamster
Clone GL3
Gene ID (Entrez) 6965
Monoclonal or Polyclonal Monoclonal
Storage Requirements Store at 4°C. Do not freeze.
Conjugate Unlabeled
Concentration 0.5mg/mL
Format Affinity Purified
Primary or Secondary Primary
Immunogen The immunogen for this antibody was gamma delta TCR.
Purification Method Affinity Purified
Isotype IgG
Antigen TCR gamma/delta
Quantity 0.5mg