Please login to your online account to display your discounted pricing

Armenian Hamster anti-TCR gamma/delta, Clone: GL3, Biotin, Novus Biologicals™

$252.00 - $725.00

Specifications

Antigen TCR gamma/delta
Clone GL3
Immunogen The immunogen for this antibody was gamma delta TCR.
Gene ID (Entrez) 6965
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14354901

 
novus biologicals canada
NBP14354901MG
0.1mg Each for $329.00

NBP14354905

 
novus biologicals canada
NBP143549005MG
0.05mg Each for $252.00

NBP1435495

 
novus biologicals canada
NBP14354905MG
0.5mg Each for $725.00
Description & Specifications

Specifications

Antigen TCR gamma/delta
Clone GL3
Immunogen The immunogen for this antibody was gamma delta TCR.
Gene ID (Entrez) 6965
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Isotype IgG
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry