Please login to your online account to display your discounted pricing

Armenian Hamster anti-TCR beta, Clone: H57-597, PE-Cy5, Novus Biologicals™

$252.00 - $505.00

Specifications

Antigen TCR beta
Clone H57-597
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was TCR beta.
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14306701

 
novus biologicals canada
NBP14306701MG
0.1mg Each for $362.00

NBP14306705

 
novus biologicals canada
NBP143067005MG
0.05mg Each for $252.00

NBP14306702

 
novus biologicals canada
NBP14306702MG
0.2mg Each for $505.00
Description & Specifications

Specifications

Antigen TCR beta
Clone H57-597
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was TCR beta.
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Mouse
Isotype IgG
Conjugate PE-Cy5
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry