Please login to your online account to display your discounted pricing

Armenian Hamster anti-TCR beta, Clone: H57-597, Novus Biologicals™

$274.00 - $362.00

Specifications

Antigen TCR beta
Clone H57-597
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was TCR beta.
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14327101

 
novus biologicals canada
NBP14327101MG
0.1mg Each for $274.00

NBP1432715

 
novus biologicals canada
NBP14327105MG
0.5mg Each for $362.00
Description & Specifications

Specifications

Antigen TCR beta
Clone H57-597
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was TCR beta.
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Mouse
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)