Please login to your online account to display your discounted pricing

Armenian Hamster anti-TCR beta, Clone: H57-597, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14327105MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1432715

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Immunogen The immunogen for this antibody was TCR beta.
Antigen TCR beta
Formulation Phosphate buffer pH 7.2
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Isotype IgG
Purification Method Affinity Purified
Quantity 0.5mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone H57-597
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Host Species Armenian Hamster
Storage Requirements Store at 4°C. Do not freeze.