Please login to your online account to display your discounted pricing

Armenian Hamster anti-TCR beta, Clone: H57-597, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14327101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14327101

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Antigen TCR beta
Host Species Armenian Hamster
Immunogen The immunogen for this antibody was TCR beta.
Isotype IgG
Quantity 0.1mg
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity Mouse
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Purification Method Affinity Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Clone H57-597
Concentration 0.5mg/mL