Please login to your online account to display your discounted pricing

Armenian Hamster anti-TCR beta, Clone: H57-597, Cy5.5, PerCP, Novus Biologicals™

$197.00 - $395.00

Specifications

Antigen TCR beta
Clone H57-597
Immunogen The immunogen for this antibody was TCR beta.
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14283303

 
novus biologicals canada
NBP1428330025MG
0.025mg Each for $197.00

NBP14283301

 
novus biologicals canada
NBP14283301MG
0.1mg Each for $395.00
Description & Specifications

Specifications

Antigen TCR beta
Clone H57-597
Immunogen The immunogen for this antibody was TCR beta.
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Mouse
Isotype IgG
Conjugate Cy5.5, PerCP
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry