Please login to your online account to display your discounted pricing

Armenian Hamster anti-STAT1, Clone: ATO-2F5, Novus Biologicals™

$252.00 - $362.00

Specifications

Antigen STAT1
Clone ATO-2F5
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was STAT1.
Gene ID (Entrez) 6772
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14326401

 
novus biologicals canada
NBP14326401MG
0.1mg Each for $362.00

NBP14326403

 
novus biologicals canada
NBP1432640025MG
0.025mg Each for $252.00
Description & Specifications

Specifications

Antigen STAT1
Clone ATO-2F5
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was STAT1.
Gene ID (Entrez) 6772
Formulation Aqueous buffer (contains stabilizer if necessary)
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Human
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Western Blotting