Please login to your online account to display your discounted pricing

Armenian Hamster anti-STAT1, Clone: ATO-2F5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1432640025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14326403

  • / Each
 

Description & Specifications

Specifications

Antigen STAT1
Formulation Aqueous buffer (contains stabilizer if necessary)
Host Species Armenian Hamster
Applications Western Blotting
Immunogen The immunogen for this antibody was STAT1.
Isotype IgG
Purification Method Affinity Purified
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6772
Cross Reactivity Human
Format Affinity Purified
Quantity 0.025mg
Storage Requirements Store at 4°C. Do not freeze.
Clone ATO-2F5
Concentration 0.5mg/mL
Conjugate Unlabeled