Please login to your online account to display your discounted pricing

Armenian Hamster anti-STAT1, Clone: ATO-2F5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14326401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14326401

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Formulation Aqueous buffer (contains stabilizer if necessary)
Host Species Armenian Hamster
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Clone ATO-2F5
Conjugate Unlabeled
Cross Reactivity Human
Antigen STAT1
Applications Western Blotting
Immunogen The immunogen for this antibody was STAT1.
Isotype IgG
Purification Method Affinity Purified
Primary or Secondary Primary
Gene ID (Entrez) 6772
Concentration 0.5mg/mL