Please login to your online account to display your discounted pricing

Armenian Hamster anti-Plexin B2, Clone: 3E7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143310005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14331005

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Host Species Armenian Hamster
Immunogen The immunogen for this antibody was PlexinB2.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 23654
Concentration 0.5mg/mL
Cross Reactivity Mouse
Antigen Plexin B2
Format Affinity Purified
Formulation PBS [pH 7.2]
Isotype IgG
Purification Method Affinity Purified
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Clone 3E7
Conjugate Unlabeled