Please login to your online account to display your discounted pricing

Armenian Hamster anti-Plexin B2, Clone: 3E7, Biotin, Novus Biologicals™

$197.00 - $725.00

Specifications

Antigen Plexin B2
Clone 3E7
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was PlexinB2.
Gene ID (Entrez) 23654
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14355101

 
novus biologicals canada
NBP14355101MG
0.1mg Each for $329.00

NBP14355105

 
novus biologicals canada
NBP143551005MG
0.05mg Each for $197.00

NBP1435515

 
novus biologicals canada
NBP14355105MG
0.5mg Each for $725.00
Description & Specifications

Specifications

Antigen Plexin B2
Clone 3E7
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was PlexinB2.
Gene ID (Entrez) 23654
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Mouse
Isotype IgG
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry