Please login to your online account to display your discounted pricing

Armenian Hamster anti-PD-1, Clone: J43, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143110005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14311005

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Primary or Secondary Primary
Host Species Armenian Hamster
Isotype IgG
Immunogen The immunogen for this antibody was PD1.
Gene ID (Entrez) 5133
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Concentration 0.2mg/mL
Cross Reactivity Mouse
Clone J43
Quantity 0.05mg
Antigen PD-1
Purification Method Affinity Purified