Please login to your online account to display your discounted pricing

Armenian Hamster anti-PD-1, Clone: J43, FITC, Novus Biologicals™

$252.00 - $725.00

Specifications

Antigen PD-1
Clone J43
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was PD1.
Gene ID (Entrez) 5133
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14391101

 
novus biologicals canada
NBP14391101MG
0.1mg Each for $329.00

NBP14391105

 
novus biologicals canada
NBP143911005MG
0.05mg Each for $252.00

NBP1439115

 
novus biologicals canada
NBP14391105MG
0.5mg Each for $725.00
Description & Specifications

Specifications

Antigen PD-1
Clone J43
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was PD1.
Gene ID (Entrez) 5133
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Mouse
Isotype IgG
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry