Please login to your online account to display your discounted pricing

Armenian Hamster anti-Mouse IgM, Clone: GL3, PE-Cy5, Novus Biologicals™

$252.00 - $362.00

Specifications

Antigen Mouse IgM
Clone GL3
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was Phycoerythringamma delta TCR.
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14306901

 
novus biologicals canada
NBP14306901MG
0.1mg Each for $362.00

NBP14306905

 
novus biologicals canada
NBP143069005MG
0.05mg Each for $252.00
Description & Specifications

Specifications

Antigen Mouse IgM
Clone GL3
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was Phycoerythringamma delta TCR.
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Mouse
Isotype IgG
Conjugate PE-Cy5
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry