Please login to your online account to display your discounted pricing

Armenian Hamster anti-LAIR1, Clone: 113, PE, Novus Biologicals™

$246.00 - $354.00

Specifications

Antigen LAIR1
Clone 113
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD305.
Gene ID (Entrez) 3903
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14378401

 
novus biologicals canada
NBP14378401MG
0.1mg Each for $354.00

NBP14378405

 
novus biologicals canada
NBP143784005MG
0.05mg Each for $246.00
Description & Specifications

Specifications

Antigen LAIR1
Clone 113
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD305.
Gene ID (Entrez) 3903
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Mouse
Isotype IgG
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry