Please login to your online account to display your discounted pricing

Armenian Hamster anti-LAIR1, Clone: 113, PE, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP14378401MG

 View all options for this product


For Research Use Only

Catalog No. NBP14378401


Description & Specifications


Antigen LAIR1
Applications Flow Cytometry
Clone 113
Concentration 0.2mg/mL
Conjugate PE
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS [pH 7.2]
Host Species Armenian Hamster
Immunogen The immunogen for this antibody was CD305.
Isotype IgG
Purification Method Affinity Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3903