Please login to your online account to display your discounted pricing

Armenian Hamster anti-Integrin beta 3/CD61, Clone: 2C9.G3, PE, Novus Biologicals™

$192.00 - $418.00

Specifications

Antigen Integrin beta 3/CD61
Clone 2C9.G3
Immunogen The immunogen for this antibody was PE CD61.
Gene ID (Entrez) 3690
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14385401

 
novus biologicals canada
NBP14385401MG
0.1mg Each for $289.00

NBP14385402

 
novus biologicals canada
NBP14385402MG
0.2mg Each for $418.00

NBP14385405

 
novus biologicals canada
NBP143854005MG
0.05mg Each for $192.00
Description & Specifications

Specifications

Antigen Integrin beta 3/CD61
Clone 2C9.G3
Immunogen The immunogen for this antibody was PE CD61.
Gene ID (Entrez) 3690
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Mouse
Isotype IgG
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry