Please login to your online account to display your discounted pricing

Armenian Hamster anti-Integrin beta 3/CD61, Clone: 2C9.G3, PE, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP14385401MG

 View all options for this product


For Research Use Only

Catalog No. NBP14385401


Description & Specifications


Antigen Integrin beta 3/CD61
Applications Flow Cytometry
Clone 2C9.G3
Conjugate PE
Cross Reactivity Mouse
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Armenian Hamster
Immunogen The immunogen for this antibody was PE CD61.
Isotype IgG
Purification Method Affinity Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3690