Please login to your online account to display your discounted pricing

Armenian Hamster anti-Integrin beta 3/CD61, Clone: 2C9.G3, PE, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143854005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14385405

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Applications Flow Cytometry
Cross Reactivity Mouse
Antigen Integrin beta 3/CD61
Immunogen The immunogen for this antibody was PE CD61.
Quantity 0.05mg
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Formulation Phosphate buffer pH 7.2
Isotype IgG
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Clone 2C9.G3
Conjugate PE
Host Species Armenian Hamster
Gene ID (Entrez) 3690