Please login to your online account to display your discounted pricing

Armenian Hamster anti-Integrin beta 3/CD61, Clone: 2C9.G3, PE, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14385402MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14385402

  • / Each
 

Description & Specifications

Specifications

Host Species Armenian Hamster
Applications Flow Cytometry
Gene ID (Entrez) 3690
Conjugate PE
Cross Reactivity Mouse
Antigen Integrin beta 3/CD61
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Immunogen The immunogen for this antibody was PE CD61.
Isotype IgG
Purification Method Affinity Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 2C9.G3