Please login to your online account to display your discounted pricing

Armenian Hamster anti-Integrin beta 1/CD29, Clone: HMb1-1, PE, Novus Biologicals™

$197.00 - $395.00

Specifications

Antigen Integrin beta 1/CD29
Clone HMb1-1
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD29.
Gene ID (Entrez) 3688
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14387401

 
novus biologicals canada
NBP14387401MG
0.1mg Each for $252.00

NBP14387402

 
novus biologicals canada
NBP14387402MG
0.2mg Each for $395.00

NBP14387405

 
novus biologicals canada
NBP143874005MG
0.05mg Each for $197.00
Description & Specifications

Specifications

Antigen Integrin beta 1/CD29
Clone HMb1-1
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD29.
Gene ID (Entrez) 3688
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Isotype IgG
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry