Please login to your online account to display your discounted pricing

Armenian Hamster anti-Integrin beta 1/CD29, Clone: HMb1-1, PE-Cy7, Novus Biologicals™

$175.00 - $362.00

Specifications

Antigen Integrin beta 1/CD29
Clone HMb1-1
Immunogen The immunogen for this antibody was CD29.
Gene ID (Entrez) 3688
Formulation PBS [pH 7.2]
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14296001

 
novus biologicals canada
NBP14296001MG
0.1mg Each for $362.00

NBP14296003

 
novus biologicals canada
NBP1429600025MG
0.025mg Each for $175.00
Description & Specifications

Specifications

Antigen Integrin beta 1/CD29
Clone HMb1-1
Immunogen The immunogen for this antibody was CD29.
Gene ID (Entrez) 3688
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Isotype IgG
Conjugate PE-Cy7
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry