Please login to your online account to display your discounted pricing

Armenian Hamster anti-Integrin beta 1/CD29, Clone: HMb1-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14342405MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1434245

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunoprecipitation
Cross Reactivity Mouse
Antigen Integrin beta 1/CD29
Format Affinity Purified
Immunogen The immunogen for this antibody was CD29.
Isotype IgG
Quantity 0.5mg
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3688
Conjugate Unlabeled
Formulation PBS [pH 7.2]
Host Species Armenian Hamster
Purification Method Affinity Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Clone HMb1-1
Concentration 0.5mg/mL