Please login to your online account to display your discounted pricing

Armenian Hamster anti-Integrin beta 1/CD29, Clone: HMb1-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143424005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14342405

  • / Each
 

Description & Specifications

Specifications

Antigen Integrin beta 1/CD29
Applications Flow Cytometry
Applications Immunoprecipitation
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS [pH 7.2]
Host Species Armenian Hamster
Isotype IgG
Purification Method Affinity Purified
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3688
Clone HMb1-1
Concentration 0.5mg/mL
Conjugate Unlabeled
Immunogen The immunogen for this antibody was CD29.