Please login to your online account to display your discounted pricing

Armenian Hamster anti-Integrin beta 1/CD29, Clone: HMb1-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14342401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14342401

  • / Each
 

Description & Specifications

Specifications

Formulation PBS [pH 7.2]
Applications Flow Cytometry
Applications Immunoprecipitation
Immunogen The immunogen for this antibody was CD29.
Gene ID (Entrez) 3688
Conjugate Unlabeled
Antigen Integrin beta 1/CD29
Host Species Armenian Hamster
Isotype IgG
Purification Method Affinity Purified
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone HMb1-1
Format Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Concentration 0.5mg/mL
Cross Reactivity Mouse