Please login to your online account to display your discounted pricing

Armenian Hamster anti-Integrin beta 1/CD29, Clone: HMb1-1, Biotin, Novus Biologicals™

$98.00 - $197.00

Specifications

Antigen Integrin beta 1/CD29
Clone HMb1-1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD29.
Gene ID (Entrez) 3688
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14365701

 
novus biologicals canada
NBP14365701MG
0.1mg Each for $197.00

NBP14365703

 
novus biologicals canada
NBP1436570025MG
0.025mg Each for $98.00
Description & Specifications

Specifications

Antigen Integrin beta 1/CD29
Clone HMb1-1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD29.
Gene ID (Entrez) 3688
Formulation Aqueous buffer (contains stabilizer if necessary)
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Mouse
Isotype IgG
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)