missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Armenian Hamster anti-Integrin beta 1/CD29, Clone: HMb1-1, APC, Novus Biologicals™

$175.00 - $362.00

Specifications

Antigen Integrin beta 1/CD29
Clone HMb1-1
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD29.
Gene ID (Entrez) 3688
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
NBP14304501 Novus Biologicals Canada
NBP14304501MG
0.1mg Each for $362.00
NBP14304503 Novus Biologicals Canada
NBP1430450025MG
0.025mg Each for $175.00
Specifications Description & Specifications

Specifications

Integrin beta 1/CD29
0.2mg/mL
3688
Monoclonal
Store at 4°C. Do not freeze.
IgG
Affinity Purified
Flow Cytometry
HMb1-1
The immunogen for this antibody was CD29.
PBS [pH 7.2]
Primary
Armenian Hamster
Allophycocyanin
Affinity Purified