Please login to your online account to display your discounted pricing

Armenian Hamster anti-Integrin alpha E/CD103, Clone: 2E7, APC, Novus Biologicals™

$171.00 - $354.00

Specifications

Antigen Integrin alpha E/CD103
Clone 2E7
Concentration 0.2mg/mL
Immunogen The CD103/alpha IELbeta7 integrin complex (Mw 160 and 115 kDa peptides).
Gene ID (Entrez) 3682
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14302401

 
novus biologicals canada
NBP14302401MG
0.1mg Each for $354.00

NBP14302403

 
novus biologicals canada
NBP1430240025MG
0.025mg Each for $171.00
Description & Specifications

Specifications

Antigen Integrin alpha E/CD103
Clone 2E7
Concentration 0.2mg/mL
Immunogen The CD103/alpha IELbeta7 integrin complex (Mw 160 and 115 kDa peptides).
Gene ID (Entrez) 3682
Formulation PBS [pH 7.2]. IgG purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Mouse
Isotype IgG
Conjugate APC
Format Purified
Purification Method Purified
Applications Flow Cytometry