missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Armenian Hamster anti-Integrin alpha 5/CD49e, Clone: HMa5-1, FITC, Novus Biologicals™

$131.00 - $329.00

Specifications

Antigen Integrin alpha 5/CD49e
Clone HMa5-1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD49e.
Gene ID (Entrez) 3678
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14400102 Novus Biologicals Canada
NBP14400102MG
0.2mg Each for $329.00
NBP14400105 Novus Biologicals Canada
NBP144001005MG
0.05mg Each for $131.00
Specifications Description & Specifications

Specifications

Integrin alpha 5/CD49e
0.5mg/mL
3678
Monoclonal
Store at 4°C. Do not freeze.
IgG
Affinity Purified
Flow Cytometry
HMa5-1
The immunogen for this antibody was CD49e.
PBS [pH 7.2]
Primary
Armenian Hamster
FITC
Affinity Purified