Please login to your online account to display your discounted pricing

Armenian Hamster anti-IL-1 alpha/IL-1F1, Clone: ALF-161, Novus Biologicals™

$274.00 - $406.00

Specifications

Antigen IL-1 alpha/IL-1F1
Clone ALF-161
Immunogen The immunogen for this antibody was IL1a.
Gene ID (Entrez) 3552
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14324705

 
novus biologicals canada
NBP143247005MG
0.05mg Each for $274.00

NBP1432475

 
novus biologicals canada
NBP14324705MG
0.5mg Each for $406.00
Description & Specifications

Specifications

Antigen IL-1 alpha/IL-1F1
Clone ALF-161
Immunogen The immunogen for this antibody was IL1a.
Gene ID (Entrez) 3552
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Mouse
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications ELISA