Please login to your online account to display your discounted pricing

Armenian Hamster anti-IL-1 alpha/IL-1F1, Clone: ALF-161, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14324705MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1432475

  • / Each
 

Description & Specifications

Specifications

Antigen IL-1 alpha/IL-1F1
Immunogen The immunogen for this antibody was IL1a.
Cross Reactivity Mouse
Quantity 0.5mg
Clone ALF-161
Applications ELISA
Host Species Armenian Hamster
Conjugate Unlabeled
Isotype IgG
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3552
Storage Requirements Store at 4°C. Do not freeze.
Purification Method Affinity Purified
Formulation Phosphate buffer pH 7.2
Regulatory Status RUO
Format Affinity Purified
Primary or Secondary Primary