Please login to your online account to display your discounted pricing

Armenian Hamster anti-IL-1 alpha/IL-1F1, Clone: ALF-161, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143247005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14324705

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Cross Reactivity Mouse
Clone ALF-161
Storage Requirements Store at 4°C. Do not freeze.
Immunogen The immunogen for this antibody was IL1a.
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Quantity 0.05mg
Antigen IL-1 alpha/IL-1F1
Host Species Armenian Hamster
Isotype IgG
Purification Method Affinity Purified
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3552
Format Affinity Purified
Formulation Phosphate buffer pH 7.2