Please login to your online account to display your discounted pricing

Armenian Hamster anti-IFN-gamma R1/CD119, Clone: 2E2, PE, Novus Biologicals™

$175.00 - $362.00

Specifications

Antigen IFN-gamma R1/CD119
Clone 2E2
Immunogen The immunogen for this antibody was CD119.
Gene ID (Entrez) 3459
Formulation PBS [pH 7.2]
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14381901

 
novus biologicals canada
NBP14381901MG
0.1mg Each for $362.00

NBP14381903

 
novus biologicals canada
NBP1438190025MG
0.025mg Each for $175.00
Description & Specifications

Specifications

Antigen IFN-gamma R1/CD119
Clone 2E2
Immunogen The immunogen for this antibody was CD119.
Gene ID (Entrez) 3459
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Mouse
Isotype IgG
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry