Please login to your online account to display your discounted pricing

Armenian Hamster anti-ICOS, Clone: C398.4A, PE-Cy5, Novus Biologicals™

$197.00 - $329.00

Specifications

Antigen ICOS
Clone C398.4A
Concentration 0.2mg/mL
Immunogen Mouse T cell clone D10.G4.1
Gene ID (Entrez) 29851
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14306501

 
novus biologicals canada
NBP14306501MG
0.1mg Each for $329.00

NBP14306505

 
novus biologicals canada
NBP143065005MG
0.05mg Each for $197.00
Description & Specifications

Specifications

Antigen ICOS
Clone C398.4A
Concentration 0.2mg/mL
Immunogen Mouse T cell clone D10.G4.1
Gene ID (Entrez) 29851
Formulation Phosphate buffer pH 7.2. Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C in the dark.
Host Species Armenian Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG
Conjugate PE-Cy5
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry