missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Armenian Hamster anti-Fc epsilon RI, Clone: MAR-1, Biotin, Novus Biologicals™

$252.00 - $725.00

Specifications

Antigen Fc epsilon RI
Clone MAR-1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was FceRI alpha.
Gene ID (Entrez) 2205
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14352301 Novus Biologicals Canada
NBP14352301MG
0.1mg Each for $329.00
NBP14352305 Novus Biologicals Canada
NBP143523005MG
0.05mg Each for $252.00
NBP1435235 Novus Biologicals Canada
NBP14352305MG
0.5mg Each for $725.00
Specifications Description & Specifications

Specifications

Fc epsilon RI
0.5mg/mL
2205
Monoclonal
Store at 4°C. Do not freeze.
IgG
Affinity Purified
Flow Cytometry
MAR-1
The immunogen for this antibody was FceRI alpha.
Phosphate buffer pH 7.2
Primary
Armenian Hamster
Biotin
Affinity Purified