Please login to your online account to display your discounted pricing

Armenian Hamster anti-Fas Ligand/TNFSF6, Clone: MFL4, Novus Biologicals™

$246.00 - $354.00

Specifications

Antigen Fas Ligand/TNFSF6
Clone MFL4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD95 Ligand.
Gene ID (Entrez) 356
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14327501

 
novus biologicals canada
NBP14327501MG
0.1mg Each for $321.00

NBP1432755

 
novus biologicals canada
NBP14327505MG
0.5mg Each for $354.00

NBP14327505

 
novus biologicals canada
NBP143275005MG
0.05mg Each for $246.00
Description & Specifications

Specifications

Antigen Fas Ligand/TNFSF6
Clone MFL4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD95 Ligand.
Gene ID (Entrez) 356
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Armenian Hamster
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Functional Assay