Please login to your online account to display your discounted pricing

Armenian Hamster anti-Fas Ligand/TNFSF6, Clone: MFL4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143275005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14327505

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Functional Assay
Primary or Secondary Primary
Formulation Phosphate buffer pH 7.2
Regulatory Status RUO
Antigen Fas Ligand/TNFSF6
Immunogen The immunogen for this antibody was CD95 Ligand.
Format Affinity Purified
Clone MFL4
Cross Reactivity Mouse
Cross Reactivity Rat
Host Species Armenian Hamster
Gene ID (Entrez) 356
Concentration 0.5mg/mL
Monoclonal or Polyclonal Monoclonal
Isotype IgG
Storage Requirements Store at 4°C. Do not freeze.
Purification Method Affinity Purified
Conjugate Unlabeled
Quantity 0.05mg