Please login to your online account to display your discounted pricing

Armenian Hamster anti-Fas Ligand/TNFSF6, Clone: MFL4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14327501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14327501

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Applications Flow Cytometry
Applications Functional Assay
Quantity 0.1mg
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 356
Antigen Fas Ligand/TNFSF6
Formulation Phosphate buffer pH 7.2
Host Species Armenian Hamster
Isotype IgG
Purification Method Affinity Purified
Regulatory Status RUO
Primary or Secondary Primary
Clone MFL4
Conjugate Unlabeled
Cross Reactivity Mouse
Cross Reactivity Rat
Immunogen The immunogen for this antibody was CD95 Ligand.
Concentration 0.5mg/mL