Please login to your online account to display your discounted pricing

Armenian Hamster anti-Fas Ligand/TNFSF6, Clone: MFL4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14327505MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1432755

  • / Each
 

Description & Specifications

Specifications

Host Species Armenian Hamster
Applications Flow Cytometry
Applications Functional Assay
Immunogen The immunogen for this antibody was CD95 Ligand.
Quantity 0.5mg
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen Fas Ligand/TNFSF6
Formulation Phosphate buffer pH 7.2
Purification Method Affinity Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 356
Clone MFL4
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Isotype IgG
Primary or Secondary Primary