Please login to your online account to display your discounted pricing

Armenian Hamster anti-Fas Ligand/TNFSF6, Clone: MFL3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143276005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14327605

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Host Species Armenian Hamster
Applications Flow Cytometry
Applications Functional Assay
Immunogen The immunogen for this antibody was CD95 Ligand.
Isotype IgG
Quantity 0.05mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 356
Clone MFL3
Antigen Fas Ligand/TNFSF6
Formulation Phosphate buffer pH 7.2
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse